siRNA Details
| Gene Symbol: MIR155HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_001458.3 | 3 | 560 | |||
| GenBank mRNA | AF402776.1 | - | LIT1586 bic |
3 | 512 | |
| CT000653.1 | - | 3 | 560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR155HG| Gene Symbol: MIR155HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_001458.3 | 3 | 560 | |||
| GenBank mRNA | AF402776.1 | - | LIT1586 bic |
3 | 512 | |
| CT000653.1 | - | 3 | 560 | |||