siRNA Details
| Gene Symbol: PWAR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_022008.1 | 1 | 3315 | |||
| GenBank mRNA | AF019618.1 | - | LIT1110 PAR5 |
1 | 3123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PWAR5| Gene Symbol: PWAR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_022008.1 | 1 | 3315 | |||
| GenBank mRNA | AF019618.1 | - | LIT1110 PAR5 |
1 | 3123 | |