siRNA Details
| Gene Symbol: ZNF205-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024166.1 | 1 | 525 | |||
| NR_024167.1 | 1 | 525 | ||||
| GenBank mRNA | BC001809.2 | - | 1 | 525 | ||
| BC001824.2 | - | 1 | 525 | |||
| BC121099.1 | - | 1 | 152 | |||
| BC121100.1 | - | 1 | 152 | |||

ZNF205-AS1
Genomic Map