siRNA Details
| Gene Symbol: ZNF205-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024166.1 | 1 | 348 | |||
| NR_024167.1 | 1 | 348 | ||||
| GenBank mRNA | BC001809.2 | - | 1 | 348 | ||
| BC001824.2 | - | 1 | 348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF205-AS1| Gene Symbol: ZNF205-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024166.1 | 1 | 348 | |||
| NR_024167.1 | 1 | 348 | ||||
| GenBank mRNA | BC001809.2 | - | 1 | 348 | ||
| BC001824.2 | - | 1 | 348 | |||