siRNA Details
Gene Symbol: ZNF205-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024166.1 | 1 | 65 | |||
NR_024167.1 | 1 | 65 | ||||
GenBank mRNA | BC001809.2 | - | 1 | 65 | ||
BC001824.2 | - | 1 | 65 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF205-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024166.1 | 1 | 65 | |||
NR_024167.1 | 1 | 65 | ||||
GenBank mRNA | BC001809.2 | - | 1 | 65 | ||
BC001824.2 | - | 1 | 65 |