siRNA Details
| Gene Symbol: KAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028402.2 | 1 | 14 | |||
| GenBank mRNA | AK023759.1 | - | 1 | 14 | ||
| AU140233.1 | - | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KAT14| Gene Symbol: KAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028402.2 | 1 | 14 | |||
| GenBank mRNA | AK023759.1 | - | 1 | 14 | ||
| AU140233.1 | - | 1 | 14 | |||