siRNA Details
| Gene Symbol: LINC00998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015442.1 | 3 | 752 | |||
| NR_024412.1 | 3 | 840 | ||||
| GenBank mRNA | BC065765.1 | - | 3 | 728 | ||
| BC107860.1 | - | 3 | 756 | |||
| BX537645.1 | - | 3 | 725 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR625886.1 | - | 1 | 1595 | Chr. 7 - 113116990 - 113118599 [-] on Build GRCh38 | |

LINC00998
Genomic Map