siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184906.1 | NP_001171835.1 | 14 | 8430 | ||
| NM_032875.2 | NP_116264.2 | 15 | 8526 | |||
| XM_005257746.3 | XP_005257803.2 | 15 | 8717 | |||
| XM_005257747.3 | XP_005257804.2 | 14 | 8498 | |||

FBXL20
Genomic Map