siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184906.1 | NP_001171835.1 | 14 | 9226 | ||
| NM_032875.2 | NP_116264.2 | 15 | 9322 | |||
| XM_005257746.3 | XP_005257803.2 | 15 | 9513 | |||
| XM_005257747.3 | XP_005257804.2 | 14 | 9294 | |||
| GenBank mRNA | AL117623.1 | - | 1 | 384 | ||
| BC042012.1 | - | 1 | 594 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ719214.1 | - | 1 | 333 | Chr. 17 - 39253150 - 39254035 [-] on Build GRCh38 | |

FBXL20
Genomic Map