siRNA Details
| Gene Symbol: FKSG29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024013.1 | 1 | 212 | |||
| GenBank mRNA | AY014273.1 | - | 1 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FKSG29| Gene Symbol: FKSG29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024013.1 | 1 | 212 | |||
| GenBank mRNA | AY014273.1 | - | 1 | 212 | ||