siRNA Details
| Gene Symbol: HNRNPDL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003249.2 | 8 | 1857 | |||
| GenBank mRNA | AB017019.1 | - | 8 | 1857 | ||
| AK057480.1 | - | 2 | 673 | |||
| AK294683.1 | - | 5 | 1095 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK011143, BC021374.

HNRNPDL
Genomic Map