siRNA Details
Gene Symbol: MALAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002819.3 | 1 | 1822 | |||
GenBank mRNA | AJ535465.1 | - | 262 | |||
BC025986.1 | - | 1 | 543 | |||
BC104662.1 | - | 1 | 394 | |||
EF177381.1 | - | 1 | 1822 | |||
FJ209305.1 | - | 1 | 539 |
