siRNA Details
| Gene Symbol: RPPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002312.1 | 1 | 267 | |||
| GenBank mRNA | CV309337.1 | - | 1 | 185 | ||
| X15624.1 | - | 1 | 266 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CV427054.1 | - | 1 | 182 | Chr. 14 - 20343096 - 20343326 [-] on Build GRCh38 | |

RPPH1
Genomic Map