siRNA Details
Gene Symbol: EP400NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003290.2 | 10 | 1437 | |||
GenBank mRNA | AK091234.1 | - | 11 | 2641 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EP400NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003290.2 | 10 | 1437 | |||
GenBank mRNA | AK091234.1 | - | 11 | 2641 |