siRNA Details
| Gene Symbol: EP400NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003290.2 | 7, 8 | 786 | |||
| GenBank mRNA | AK091234.1 | - | 8, 9 | 1978 | ||
| AK126212.1 | - | 7, 8 | 2034 | |||
| AK128208.1 | - | 3, 4 | 1367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EP400NL| Gene Symbol: EP400NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003290.2 | 7, 8 | 786 | |||
| GenBank mRNA | AK091234.1 | - | 8, 9 | 1978 | ||
| AK126212.1 | - | 7, 8 | 2034 | |||
| AK128208.1 | - | 3, 4 | 1367 | |||