siRNA Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002196.2 | 1 | 342 | |||
| NR_131223.1 | 1 | 342 | ||||
| GenBank mRNA | AL546946.3 | - | 1 | 302 | ||
| AL548405.3 | - | 1 | 302 | |||
| DA443982.1 | - | 1 | 342 | |||
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC002550.1 | - | 132 | |||

H19
Genomic Map