siRNA Details
| Gene Symbol: NEAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028272.1 | 1 | 1153 | |||
| NR_131012.1 | 1 | 1153 | ||||
| GenBank mRNA | AF001892.1 | - | 1 | 413 | ||
| EF177379.1 | - | 1 | 1153 | |||
| GQ859162.1 | - | 1 | 1153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEAT1| Gene Symbol: NEAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028272.1 | 1 | 1153 | |||
| NR_131012.1 | 1 | 1153 | ||||
| GenBank mRNA | AF001892.1 | - | 1 | 413 | ||
| EF177379.1 | - | 1 | 1153 | |||
| GQ859162.1 | - | 1 | 1153 | |||