siRNA Details
| Gene Symbol: NEAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028272.1 | 1 | 2327 | |||
| NR_131012.1 | 1 | 2327 | ||||
| GenBank mRNA | AF001893.1 | - | 1 | 79 | ||
| AK027191.1 | - | 1 | 311 | |||
| EF177379.1 | - | 1 | 2327 | |||
| GQ859162.1 | - | 1 | 2327 | |||

NEAT1
Genomic Map