siRNA Details
| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001240.3 | NP_001231.2 | 9 | 5811 | ||
| NM_001277842.1 | NP_001264771.1 | 8 | 5647 | |||
| XM_017020197.1 | XP_016875686.1 | 8 | 5322 | |||
| GenBank mRNA | AK002085.1 | - | 1 | 1045 | ||
| AL832083.1 | - | 1 | 2445 | |||
| BX537514.1 | - | 1 | 2782 | |||
| BX537516.1 | - | 1 | 2657 | |||

CCNT1
Genomic Map