siRNA Details
| Gene Symbol: PVT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003367.3 | 7 | 1347 | |||
| GenBank mRNA | BC041065.1 | - | 6 | 1106 | ||
| BX648921.1 | - | 1 | 461 | |||
| D79458.1 | - | 4 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PVT1| Gene Symbol: PVT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003367.3 | 7 | 1347 | |||
| GenBank mRNA | BC041065.1 | - | 6 | 1106 | ||
| BX648921.1 | - | 1 | 461 | |||
| D79458.1 | - | 4 | 346 | |||