siRNA Details
| Gene Symbol: PVT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003367.3 | 9 | 1922 | |||
| GenBank mRNA | BC041065.1 | - | 8 | 1681 | ||
| BX648921.1 | - | 1 | 5160 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG184938.1 | - | 3 | 510 | Chr. 8 - 128070158 - 128101256 [+] on Build GRCh38 | |

PVT1
Genomic Map