siRNA Details
| Gene Symbol: UCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_015379.3 | 3 | 535 | |||
| GenBank mRNA | AK024303.1 | - | 2 | 715 | ||
| AK092401.1 | - | 3 | 521 | |||
| BC005351.1 | - | 3 | 387 | |||
| BC017567.1 | - | 3 | 391 | |||
| BC107722.1 | - | 3 | 391 | |||
| DQ343132.1 | - | LIT3336 UCA1 |
3 | 535 | ||
| EU334869.1 | - | 3 | 536 | |||
| GU357550.1 | - | 3 | 387 | |||
| GU799561.1 | - | 3 | 535 | |||
| GU799562.1 | - | 3 | 535 | |||
| GU799563.1 | - | 3 | 535 | |||
| GU799564.1 | - | 3 | 535 | |||
| GU799565.1 | - | 3 | 535 | |||
| HQ833208.1 | - | 3 | 535 | |||
| HQ833209.1 | - | 3 | 535 | |||

UCA1
Genomic Map