siRNA Details
| Gene Symbol: UCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_015379.3 | 1 | 68 | |||
| GenBank mRNA | AK092401.1 | - | 1 | 54 | ||
| DQ343132.1 | - | LIT3336 UCA1 |
1 | 68 | ||
| EU334869.1 | - | 1 | 69 | |||
| GU799561.1 | - | 1 | 68 | |||
| GU799562.1 | - | 1 | 68 | |||
| GU799563.1 | - | 1 | 68 | |||
| GU799564.1 | - | 1 | 68 | |||
| GU799565.1 | - | 1 | 68 | |||
| HQ833208.1 | - | 1 | 68 | |||
| HQ833209.1 | - | 1 | 68 | |||

UCA1
Genomic Map