siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 3 | 3783 | |||
| NR_110492.1 | 4 | 3839 | ||||
| NR_110493.1 | 3 | 3769 | ||||
| GenBank mRNA | AB073661.1 | - | 1 | 237 | ||
| AK000749.1 | - | 4 | 1684 | |||
| AK057695.1 | - | 2 | 1807 | |||

TUG1
Genomic Map