siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 3 | 5921 | |||
| NR_110492.1 | 4 | 5977 | ||||
| NR_110493.1 | 3 | 5907 | ||||
| GenBank mRNA | AB073661.1 | - | 1 | 2376 | ||
| BM461799.1 | - | 1 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUG1| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 3 | 5921 | |||
| NR_110492.1 | 4 | 5977 | ||||
| NR_110493.1 | 3 | 5907 | ||||
| GenBank mRNA | AB073661.1 | - | 1 | 2376 | ||
| BM461799.1 | - | 1 | 137 | |||