siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 3 | 4625 | |||
| NR_110492.1 | 4 | 4681 | ||||
| NR_110493.1 | 3 | 4611 | ||||
| GenBank mRNA | AB073661.1 | - | 1 | 1080 | ||
| BM719085.1 | - | 1 | 371 | |||
| BU540086.1 | - | 1 | 197 | |||

TUG1
Genomic Map