siRNA Details
| Gene Symbol: NOL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256394.1 | NP_001243323.1 | 4 | 573 | ||
| XM_006717169.3 | XP_006717232.1 | 4 | 488 | |||
| XM_006717170.3 | XP_006717233.1 | 4 | 356 | |||
| XM_006717173.3 | XP_006717236.1 | 4 | 356 | |||
| XM_017014877.1 | XP_016870366.1 | 4 | 356 | |||
| GenBank mRNA | BC146810.1 | - | 4 | 329 | 8860369 | |

NOL8
Genomic Map