siRNA Details
| Gene Symbol: NOL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256394.1 | NP_001243323.1 | 1, 2 | 273 | ||
| XM_006717167.3 | XP_006717230.1 | 1, 2 | 273 | |||
| XM_017014879.1 | XP_016870368.1 | 1, 2 | 14 | |||
| GenBank mRNA | BC146810.1 | - | 1, 2 | 29 | 8860369 | |
| DA470513.1 | - | 1, 2 | 275 | |||

NOL8
Genomic Map