siRNA Details
| Gene Symbol: RPL19P12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026660.1 | 1 | 248 | |||
| GenBank mRNA | AW373666.1 | - | 1 | 250 | ||
| CD709306.1 | - | 1 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL19P12| Gene Symbol: RPL19P12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026660.1 | 1 | 248 | |||
| GenBank mRNA | AW373666.1 | - | 1 | 250 | ||
| CD709306.1 | - | 1 | 251 | |||