siRNA Details
| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024408.1 | 2 | 536 | |||
| GenBank mRNA | AK096706.1 | - | 1 | 2065 | ||
| AL117474.1 | - | 1 | 260 | |||
| BC028086.1 | - | 2 | 478 | |||
| BC040969.1 | - | 1 | 2011 | |||
| BX648532.1 | - | 1 | 2049 | |||

PSMD5-AS1
Genomic Map