siRNA Details
| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024408.1 | 4 | 3637 | |||
| GenBank mRNA | AL117474.1 | - | 3 | 3373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMD5-AS1| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024408.1 | 4 | 3637 | |||
| GenBank mRNA | AL117474.1 | - | 3 | 3373 | ||