siRNA Details
| Gene Symbol: PP7080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024158.1 | 2 | 1592 | |||
| GenBank mRNA | AF370427.1 | - | 2 | 1693 | ||
| BC045808.1 | - | 2 | 1431 | |||
| BC086861.1 | - | 2 | 1573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PP7080| Gene Symbol: PP7080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024158.1 | 2 | 1592 | |||
| GenBank mRNA | AF370427.1 | - | 2 | 1693 | ||
| BC045808.1 | - | 2 | 1431 | |||
| BC086861.1 | - | 2 | 1573 | |||