siRNA Details
| Gene Symbol: PP7080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024158.1 | 2 | 1374 | |||
| GenBank mRNA | AF370427.1 | - | 2 | 1475 | ||
| BC045808.1 | - | 2 | 1213 | |||
| BC072677.1 | - | 2 | 1228 | |||
| BC086861.1 | - | 2 | 1355 | |||
| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023036.1 | - | 2 | 2125 | ||

PP7080
Genomic Map