siRNA Details
| Gene Symbol: NDUFB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135998.2 | NP_001129470.1 | 1 | 330 | ||
| NM_019056.6 | NP_061929.2 | 1 | 330 | |||
| GenBank mRNA | AA287166.1 | - | 1 | 299 | ||
| AF251063.1 | - | 1 | 162 | |||
| AY359056.1 | - | 1 | 330 | |||
| BC010665.1 | - | 1 | 138 | 3870660 | ||

NDUFB11
Genomic Map