siRNA Details
| Gene Symbol: FIRRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026975.1 | 10 | 1817 | |||
| GenBank mRNA | BC131554.1 | - | 10 | 1273 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE544172.1 | - | 1 | 30 | Chr. X - 131702978 - 131749448 [-] on Build GRCh38 | |

FIRRE
Genomic Map