siRNA Details
| Gene Symbol: OIP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026757.1 | 4 | 1860 | |||
| GenBank mRNA | AY007106.1 | - | 4 | 1842 | ||
| BC037835.1 | - | 4 | 1888 | |||
| BX647764.1 | - | 4 | 1859 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129752.1 | - | 2879 | Chr. - - [-] on Build | ||

OIP5-AS1
Genomic Map