siRNA Details
| Gene Symbol: Dleu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF380425.1 | - | LIT3421 DLEU2 |
1 | 84 | |
| AK029956.1 | - | 1 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dleu2| Gene Symbol: Dleu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF380425.1 | - | LIT3421 DLEU2 |
1 | 84 | |
| AK029956.1 | - | 1 | 110 | |||