siRNA Details
| Gene Symbol: Vis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U09772.1 | - | LIT1766 His1 |
2 | 326 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U56439.1 | - | 1 | 2757 | Chr. - - [-] on Build | |

Vis1
Genomic Map