siRNA Details
| Gene Symbol: 2010103J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK008306.1 | - | LIT3439 2010103J01Rik |
2 | 290 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010103J01Rik| Gene Symbol: 2010103J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK008306.1 | - | LIT3439 2010103J01Rik |
2 | 290 | |