siRNA Details
| Gene Symbol: Hoxa11os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U20368.1 | - | LIT1566 Hoxa11as |
1 | 309 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxa11os| Gene Symbol: Hoxa11os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | U20368.1 | - | LIT1566 Hoxa11as |
1 | 309 | |