siRNA Details
| Gene Symbol: Miat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_003718.2 | 7 | 6887 | |||
| NR_033657.1 | 7 | 6931 | ||||
| GenBank mRNA | AB300594.1 | - | 7 | 6874 | ||
| AK028326.1 | - | LIT3370 RNCR2 |
1 | 1226 | ||

Miat
Genomic Map