siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AB063319.1 | - | LIT1623 Rian |
2 | 4743 | |
| AK032566.1 | - | 1 | 1359 | |||
| AK163850.1 | - | 1 | 43 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rian| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AB063319.1 | - | LIT1623 Rian |
2 | 4743 | |
| AK032566.1 | - | 1 | 1359 | |||
| AK163850.1 | - | 1 | 43 | |||