siRNA Details
| Gene Symbol: 1810013L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK013883.1 | - | LIT3366 3010002C02 |
1 | 503 | |
| AK048963.1 | - | 1 | 880 | |||
| AK079605.1 | - | 1 | 632 | |||
| AK135924.1 | - | 1 | 3592 | |||
| AK163088.1 | - | 1 | 1098 | |||

1810013L24Rik
Genomic Map