siRNA Details
| Gene Symbol: 2610100L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033490.1 | 3 | 785 | |||
| GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
3 | 760 | |
| AK163433.1 | - | 3 | 658 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX524909.1 | - | 3 | 498 | Chr. 3 - 17789957 - 17799531 [+] on Build GRCm38 | |

2610100L16Rik
Genomic Map