siRNA Details
| Gene Symbol: 2610100L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033490.1 | 1 | 229 | |||
| GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
1 | 204 | |
| AK163433.1 | - | 1 | 102 | |||
| BE647934.1 | - | 1 | 229 | |||

2610100L16Rik
Genomic Map