siRNA Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK020989.1 | - | LIT1001 Ftx |
1, 2 | 71 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727089.1 | - | 1, 2 | 69 | Chr. X - 103607709 - 103622500 [-] on Build GRCm38 | |

Ftx
Genomic Map