siRNA Details
| Gene Symbol: Gm14342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK045142.1 | - | 74 | |||
| DQ860415.1 | - | LIT3343 HAR1R |
122 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB643374.1 | - | 1 | 72 | Chr. 2 - 180831150 - 180889662 [-] on Build GRCm38 | |

Gm14342
Genomic Map