siRNA Details
| Gene Symbol: Gm14342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | DQ860415.1 | - | LIT3343 HAR1R |
21 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14342| Gene Symbol: Gm14342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | DQ860415.1 | - | LIT3343 HAR1R |
21 | ||