siRNA Details
| Gene Symbol: 2810037O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK012876.1 | - | LIT3368 2810037O22 |
1 | 1465 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810037O22Rik| Gene Symbol: 2810037O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK012876.1 | - | LIT3368 2810037O22 |
1 | 1465 | |