siRNA Details
| Gene Symbol: Mkrn1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF494488.1 | - | LIT1345 Makorin1-p1 |
1 | 60 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn1-ps1| Gene Symbol: Mkrn1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF494488.1 | - | LIT1345 Makorin1-p1 |
1 | 60 | |